1-Cyanpiperidin 95%
Lieferant: Apollo Scientific
Synonyme:
1-Cyanopiperidine, Pentamethylenecyanamide, Piperidine-1-carbonitrile
OR922869-25G
OR922869-5G
OR922869-1G
APOSOR922869-25GEA
584
EUR
APOSOR922869-25G
APOSOR922869-5G
APOSOR922869-1G
1-Cyanpiperidin 95%
1-Cyanpiperidin
Formel:
C₆H₁₀N₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006477 CAS-Nummer: 1530-87-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...