2-(2-Methoxyethyl)pyridine 95%
Lieferant: Apollo Scientific
OR922880-1G
OR922880-25G
OR922880-5G
APOSOR922880-1GEA
145
EUR
APOSOR922880-1G
APOSOR922880-25G
APOSOR922880-5G
2-(2-Methoxyethyl)pyridine 95%
2-(2-Methoxyethyl)pyridine
Formel:
C₈H₁₁NO Lagertemperatur: Raumtemperatur |
MDL:
MFCD00023547 CAS-Nummer: 114-91-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...