Methyl-2-(hydroxymethyl)acrylat 95%
Lieferant: Apollo Scientific
Synonyme:
Methyl 2-(hydroxymethyl)acrylate
OR922936-100G
OR922936-10G
OR922936-25G
APOSOR922936-100GEA
806
EUR
APOSOR922936-100G
APOSOR922936-10G
APOSOR922936-25G
Methyl-2-(hydroxymethyl)acrylat 95%
Methyl-2-(hydroxymethyl)acrylate
Formel:
C₅H₈O₃ Molecular Weight: 116,1152 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12545894 CAS-Nummer: 15484-46-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...