Phenyl Isobutyrate 95%

Lieferant: Apollo Scientific

Synonyme: Phenyl isobutyrate, Isobutyric Acid Phenyl Ester

OR922953-25G
APOSOR922953-25GEA 134 EUR
APOSOR922953-25G
Phenyl Isobutyrate 95%
Phenyl Isobutyrate
Formel: C₁₀H₁₂O₂
Molecular Weight: 164,20 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00157315
CAS-Nummer: 20279-29-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR