2-(2-Methylallyloxy)benzaldehyde 95%
Lieferant: Apollo Scientific
Synonyme:
2-((2-Methylallyl)oxy)benzaldehyde
OR922954-1G
OR922954-5G
APOSOR922954-1GEA
48
EUR
APOSOR922954-1G
APOSOR922954-5G
2-(2-Methylallyloxy)benzaldehyde 95%
2-(2-Methylallyloxy)benzaldehyde
Formel:
C₁₁H₁₂O₂ Molecular Weight: 176,22 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06751841 CAS-Nummer: 38002-87-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...