Isopropyl-Methyl-Sulfon 95%

Lieferant: Apollo Scientific

OR922988-1G OR922988-25G OR922988-5G
APOSOR922988-1GEA 125 EUR
APOSOR922988-1G APOSOR922988-25G APOSOR922988-5G
Isopropyl-Methyl-Sulfon 95%
Isopropyl methyl sulphone
Formel: C₄H₁₀O₂S
Molecular Weight: 122,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00050415
CAS-Nummer: 4853-74-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR