5-Isopropyl-1,3,4-oxadiazol-2-amine 95%

Lieferant: Apollo Scientific

OR923013-1G OR923013-5G
APOSOR923013-1GEA 253 EUR
APOSOR923013-1G APOSOR923013-5G
5-Isopropyl-1,3,4-oxadiazol-2-amine 95%
5-Isopropyl-1,3,4-oxadiazol-2-amine
Formel: C₅H₉N₃O
Molecular Weight: 127.15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09042012
CAS-Nummer: 65283-97-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR