7-Hydroxychromon 95%
Lieferant: Apollo Scientific
Synonyme:
7-Hydroxy-4-chromone, 7-Hydroxy-4H-chromen-4-one
OR923027-1G
OR923027-5G
APOSOR923027-1GEA
66.7
EUR
APOSOR923027-1G
APOSOR923027-5G
7-Hydroxychromon 95%
7-Hydroxychromon
Formel:
C₉H₆O₃ Molecular Weight: 162,1421 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00209371 CAS-Nummer: 59887-89-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...