5-(4-Chlorobutyl)hydantoin 95%

Lieferant: Apollo Scientific

OR923059-1G OR923059-5G
APOSOR923059-1GEA 48 EUR
APOSOR923059-1G APOSOR923059-5G
5-(4-Chlorobutyl)hydantoin 95%
5-(4-Chlorobutyl)hydantoin
Formel: C₇H₁₁ClN₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00059722
CAS-Nummer: 40126-55-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR