4-Methyl-1,2,3,6-tetrahydropyridine hydrochloride 95%
Lieferant: Apollo Scientific
OR923083-1G
OR923083-250MG
APOSOR923083-1GEA
149
EUR
APOSOR923083-1G
APOSOR923083-250MG
4-Methyl-1,2,3,6-tetrahydropyridine hydrochloride 95%
4-Methyl-1,2,3,6-tetrahydropyridine hydrochloride
Formel:
C₆H₁₂ClN Lagertemperatur: Raumtemperatur |
MDL:
MFCD22123337 CAS-Nummer: 95019-16-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...