Fumaroylchlorid 95%

Lieferant: Apollo Scientific

Synonyme: Fumaryl chloride

OR923090-100G OR923090-25G
APOSOR923090-100GEA 270 EUR
APOSOR923090-100G APOSOR923090-25G
Fumaroylchlorid 95%
Fumaroylchlorid
Formel: C₄H₂Cl₂O₂
Molecular Weight: 152,96 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00000733
CAS-Nummer: 627-63-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR