1H-Pyrrolo[2,3-b]pyridin-3-ylmethanol 95%
Lieferant: Apollo Scientific
Synonyme:
(1H-Pyrrolo[2,3-b]pyridin-3-yl)methanol
OR923206-1G
OR923206-250MG
APOSOR923206-1GEA
109
EUR
APOSOR923206-1G
APOSOR923206-250MG
1H-Pyrrolo[2,3-b]pyridin-3-ylmethanol 95%
1H-Pyrrolo[2,3-b]pyridin-3-ylmethanol
Formel:
C₈H₈N₂O Molecular Weight: 148,17 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11054203 CAS-Nummer: 1065100-83-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...