1-Ethyl-1-(o-tolyl)hydrazine 95%

Lieferant: Apollo Scientific

OR923232-1G OR923232-5G
APOSOR923232-1GEA 168 EUR
APOSOR923232-1G APOSOR923232-5G
1-Ethyl-1-(o-tolyl)hydrazine 95%
1-Ethyl-1-(o-tolyl)hydrazine
Formel: C₉H₁₄N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD03844791
CAS-Nummer: 910041-10-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR