1-(4-Methyl-1-naphthyl)ethanon 95%
Lieferant: Apollo Scientific
Synonyme:
1-Acetyl-4-methylnaphthalene, 1-(4-Methylnaphthalen-1-yl)ethanone
OR923390-25G
OR923390-5G
APOSOR923390-25GEA
289
EUR
APOSOR923390-25G
APOSOR923390-5G
1-(4-Methyl-1-naphthyl)ethanon 95%
1-(4-Methyl-1-naphthyl)ethanon
Formel:
C₁₃H₁₂O Molecular Weight: 184,24 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00671556 CAS-Nummer: 28418-86-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...