5-Methyl-2-hexanone oxime 95%
Lieferant: Apollo Scientific
Synonyme:
Isoamyl Methyl Ketoxime
OR923437-25G
OR923437-5G
APOSOR923437-25GEA
422
EUR
APOSOR923437-25G
APOSOR923437-5G
5-Methyl-2-hexanone oxime 95%
5-Methyl-2-hexanone oxime
Formel:
C₇H₁₅NO Molecular Weight: 129,20 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00013934 CAS-Nummer: 624-44-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...