4'-Aminopropiophenon 95%

Lieferant: Apollo Scientific

Synonyme: p-Aminopropiophenon

OR923448-5G
APOSOR923448-5GEA 238 EUR
APOSOR923448-5G
4'-Aminopropiophenon 95%
4'-Aminopropiophenon
Formel: C₉H₁₁NO
Molecular Weight: 149,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00017113
CAS-Nummer: 70-69-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR