Ethyl-o-tolylacetat 97%

Lieferant: Apollo Scientific

Synonyme: o-Tolylessigsäureethylester, (2-Methylphenyl)essigsäureethylester

OR923556-25G
APOSOR923556-25GEA 88.6 EUR
APOSOR923556-25G
Ethyl-o-tolylacetat 97%
Ethyl-o-tolylacetat
Formel: (C₆H₄CH₃)CH₂COOCH₂CH₃
Molecular Weight: 178,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00009179
CAS-Nummer: 40291-39-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR