3-Anisoylchlorid 95%

Lieferant: Apollo Scientific

Synonyme: 3-Anissäurechlorid, 3-Methoxybenzoylchlorid, 3-Methoxybenzoesäurechlorid, m-Anissäurechlorid

OR923787-100G OR923787-25G
APOSOR923787-100GEA 248 EUR
APOSOR923787-100G APOSOR923787-25G
3-Anisoylchlorid 95%
3-Anisoylchlorid
Formel: C₈H₇ClO₂
Molecular Weight: 170,6 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00000673
CAS-Nummer: 1711-05-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR