1-Phenyl-1-hexanol 95%
Lieferant: Apollo Scientific
Synonyme:
alpha-Pentylbenzyl Alcohol
OR923820-25G
OR923820-5G
APOSOR923820-25GEA
229
EUR
APOSOR923820-25G
APOSOR923820-5G
1-Phenyl-1-hexanol 95%
1-Phenyl-1-hexanol
Formel:
C₁₂H₁₈O Molecular Weight: 178,28 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02258728 CAS-Nummer: 4471-05-0 |
Learn more
![](/m-de.vwr.com/de_DE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...