Ethylenglykol Mono-P-Tolylether 95%

Lieferant: Apollo Scientific

Synonyme: 2-(p-Tolyloxy)ethanol, 4-(2-Hydroxyethoxy)toluene, 2-(4-Methylphenoxy)ethanol

OR923838-1G OR923838-5G
APOSOR923838-1GEA 26.1 EUR
APOSOR923838-1G APOSOR923838-5G
Ethylenglykol Mono-P-Tolylether 95%
Ethylene glycol mono-p-tolyl ether
Formel: C₉H₁₂O₂
Molecular Weight: 152,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00020600
CAS-Nummer: 15149-10-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR