1-(2-Naphthyl)ethanol 95%
Lieferant: Apollo Scientific
Synonyme:
alpha-Methyl-2-naphthalenemethanol, 2-(1-Hydroxyethyl)naphthalene
OR923927-25G
APOSOR923927-25GEA
81.2
EUR
APOSOR923927-25G
1-(2-Naphthyl)ethanol 95%
1-(2-Naphthyl)ethanol
Formel:
C₁₂H₁₂O Molecular Weight: 172,23 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004111 CAS-Nummer: 7228-47-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...