Methyl-4-methoxy-2-methylbenzoate 95%

Lieferant: Apollo Scientific

Synonyme: Methyl 4-methoxy-2-methylbenzoate

OR924086-5G
APOSOR924086-5GEA 101 EUR
APOSOR924086-5G
Methyl-4-methoxy-2-methylbenzoate 95%
Methyl-4-methoxy-2-methylbenzoate
Formel: C₁₀H₁₂O₃
Molecular Weight: 180,21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06203918
CAS-Nummer: 35598-05-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR