(4-Methyl-3-pyridinyl)methanamine 95%

Lieferant: Apollo Scientific

Synonyme: (4-Methylpyridin-3-yl)methanamine

OR924116-1G OR924116-250MG
APOSOR924116-1GEA 362 EUR
APOSOR924116-1G APOSOR924116-250MG
(4-Methyl-3-pyridinyl)methanamine 95%
(4-Methyl-3-pyridinyl)methanamine
Formel: C₇H₁₀N₂
Molecular Weight: 122,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06213859
CAS-Nummer: 1443-42-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR