sec-Butyl acetoacetate 95%
Lieferant: Apollo Scientific
Synonyme:
Acetoacetic Acid sec-Butyl Ester
OR924151-25G
OR924151-5G
APOSOR924151-25GEA
377
EUR
APOSOR924151-25G
APOSOR924151-5G
sec-Butyl acetoacetate 95%
sec-Butyl acetoacetate
Formel:
C₈H₁₄O₃ Molecular Weight: 158,20 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00059392 CAS-Nummer: 13562-76-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...