Isobutylaminhydrochlorid 98%
Lieferant: Apollo Scientific
Synonyme:
2-Methylpropan-1-amine hydrochloride, 1-Amino-2-methylpropane Hydrochloride
OR924170-25G
OR924170-100G
APOSOR924170-25GEA
79.9
EUR
APOSOR924170-25G
APOSOR924170-100G
Isobutylaminhydrochlorid 98%
Isobutylamine hydrochloride
Formel:
C₄H₁₁N Molecular Weight: 109,60 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00060199 CAS-Nummer: 5041-09-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...