4-Isobutoxy-2-butanon 95%

Lieferant: Apollo Scientific

OR924228-10G OR924228-1G OR924228-5G
APOSOR924228-10GEA 609 EUR
APOSOR924228-10G APOSOR924228-1G APOSOR924228-5G
4-Isobutoxy-2-butanon 95%
4-Isobutoxy-2-butanon
Formel: C₈H₁₆O₂
Molecular Weight: 144,21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00059348
CAS-Nummer: 31576-33-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR