3-Hydroxy-3-methylpentansäure 95%

Lieferant: Apollo Scientific

Synonyme: 3-Hydroxy-3-methylpentanoic Acid

OR924447-10G OR924447-1G OR924447-5G
APOSOR924447-10GEA 493 EUR
APOSOR924447-10G APOSOR924447-1G APOSOR924447-5G
3-Hydroxy-3-methylpentansäure 95%
3-Hydroxy-3-methylpentansäure
Formel: C₆H₁₂O₃
Molecular Weight: 132,16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00059657
CAS-Nummer: 150-96-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR