N-Formyl-Dl-Phenylalanin 95%
Lieferant: Apollo Scientific
Synonyme:
2-Formamido-3-phenylpropanoic acid
OR924498-25G
OR924498-5G
APOSOR924498-25GEA
192
EUR
APOSOR924498-25G
APOSOR924498-5G
N-Formyl-Dl-Phenylalanin 95%
N-Formyl-Dl-Phenylalanin
Formel:
C₁₀H₁₁NO₃ Molecular Weight: 193,20 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00063281 CAS-Nummer: 4289-95-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...