2,2'-Bis(2-oxazoline) 95%
Lieferant: Apollo Scientific
OR924511-5G
APOSOR924511-5GEA
163
EUR
APOSOR924511-5G
2,2'-Bis(2-oxazoline) 95%
2,2'-Bis(2-oxazoline)
Formel:
C₆H₈N₂O₂ Molecular Weight: 140,14 g/mol Schmelzpunkt: 213 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00059930 CAS-Nummer: 36697-72-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...