4-tert-Butylpyridin 95%
Lieferant: Apollo Scientific
Synonyme:
4-(tert-Butyl)pyridine
OR924555-25G
APOSOR924555-25GEA
59.6
EUR
APOSOR924555-25G
4-tert-Butylpyridin 95%
4-tert-Butylpyridin
Formel:
C₉H₁₃N Molecular Weight: 135,21 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006435 CAS-Nummer: 3978-81-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...