Butyl oxamate 95%
Lieferant: Apollo Scientific
Synonyme:
Oxamic Acid Butyl Ester, Oxamidic Acid Butyl Ester
OR924585-25G
OR924585-5G
APOSOR924585-25GEA
248
EUR
APOSOR924585-25G
APOSOR924585-5G
Butyl oxamate 95%
Butyl oxamate
Formel:
C₆H₁₁NO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00059092 CAS-Nummer: 585-28-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...