Maltolpropionat 95%

Lieferant: Apollo Scientific

Synonyme: 2-Methyl-4-oxo-4H-pyran-3-yl Propionate, 2-Methyl-4-oxo-4H-pyran-3-yl propionate, 3-Hydroxy-2-methyl-4-pyrone Propionate

OR924628-25G OR924628-5G
APOSOR924628-25GEA 106 EUR
APOSOR924628-25G APOSOR924628-5G
Maltolpropionat 95%
Maltolpropionat
Formel: C₉H₁₀O₄
Molecular Weight: 182,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00036605
CAS-Nummer: 68555-63-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR