5-Phenylhydantoin 95%
Lieferant: Apollo Scientific
Synonyme:
5-Phenylimidazolidine-2,4-dione
OR924678-5G
APOSOR924678-5GEA
197
EUR
APOSOR924678-5G
5-Phenylhydantoin 95%
5-Phenylhydantoin
Formel:
C₉H₈N₂O₂ Molecular Weight: 176,1720 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00037881 CAS-Nummer: 89-24-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...