3-(2-Chlorphenyl)propiononitril 95%
Lieferant: Apollo Scientific
Synonyme:
3-(2-Chlorophenyl)propanenitrile
OR924686-5G
APOSOR924686-5GEA
101
EUR
APOSOR924686-5G
3-(2-Chlorphenyl)propiononitril 95%
3-(2-Chlorphenyl)propiononitril
Formel:
C₉H₈ClN Molecular Weight: 165,6200 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00019873 CAS-Nummer: 7315-17-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...