4-Methylcyclohexanamin (cis/trans-Gemisch)

Lieferant: Apollo Scientific

Synonyme: 1-Amino-4-methylcyclohexane

OR924724-1G OR924724-25G OR924724-5G
APOSOR924724-1GEA 29 EUR
APOSOR924724-1G APOSOR924724-25G APOSOR924724-5G
4-Methylcyclohexanamin (cis/trans-Gemisch)
4-Methylcyclohexylamine (cis- and trans- mixture)
Formel: C₇H₁₅N
Molecular Weight: 113.20 g/mol
Siedepunkt: 148…154 °C
Dichte: 0.855 g/cm³ (25 °C)
Flammpunkt: 27 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00001495
CAS-Nummer: 6321-23-9
UN: 2734
ADR: 8+3 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR