Methyl-2-ethoxybenzoat 95%
Lieferant: Apollo Scientific
Synonyme:
Methyl 2-ethoxybenzoate
OR924750-25G
APOSOR924750-25GEA
55.1
EUR
APOSOR924750-25G
Methyl-2-ethoxybenzoat 95%
Methyl-2-ethoxybenzoat
Formel:
C₁₀H₁₂O₃ Molecular Weight: 180,2005 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00017261 CAS-Nummer: 3686-55-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...