3-(1-Piperidinylmethyl)phenol 95%

Lieferant: Apollo Scientific

Synonyme: 3-(Piperidin-1-ylmethyl)phenol

OR924807-25G
APOSOR924807-25GEA 173 EUR
APOSOR924807-25G
3-(1-Piperidinylmethyl)phenol 95%
3-(1-Piperidinylmethyl)phenol
Formel: C₁₂H₁₇NO
Molecular Weight: 191.27 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00800250
CAS-Nummer: 73279-04-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR