Benzoyl-β-alanine 98%

Lieferant: Apollo Scientific

Synonyme: Betamipron, 3-Benzamidopropanoic acid

OR924812-25G OR924812-100G
APOSOR924812-25GEA 45.1 EUR
APOSOR924812-25G APOSOR924812-100G
Benzoyl-β-alanine 98%
Benzoyl-beta-alanine
Formel: C₁₀H₁₁NO₃
Molecular Weight: 193.20 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00037293
CAS-Nummer: 3440-28-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR