1-Vinyl-2-pyrrolidon 98%

Lieferant: Apollo Scientific

Synonyme: N-Vinyl-2-pyrrolidone, 1-Vinyl-2-pyrrolidinone, N-Vinyl-2-pyrrolidinone

OR924885-100G OR924885-25G OR924885-5G
APOSOR924885-100GEA 71.2 EUR
APOSOR924885-100G APOSOR924885-25G APOSOR924885-5G
1-Vinyl-2-pyrrolidon 98%
1-Vinyl-2-pyrrolidon
Formel: C₆H₉NO
Molecular Weight: 111,1418 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00003197
CAS-Nummer: 88-12-0
UN: 2810
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR