3,3'-Oxydipropannitril 98%

Lieferant: Apollo Scientific

Synonyme: Bis(2-cyanoethyl) Ether

OR924961-1G OR924961-5G
APOSOR924961-1GEA 190 EUR
APOSOR924961-1G APOSOR924961-5G
3,3'-Oxydipropannitril 98%
3,3'-Oxydipropannitril
Formel: C₆H₈N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00001960
CAS-Nummer: 1656-48-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR