1-Ethyl-1H-imidazole-2-carbaldehyde 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Ethylimidazole-2-carboxaldehyde

OR924988-10G OR924988-1G OR924988-5G
APOSOR924988-10GEA 187 EUR
APOSOR924988-10G APOSOR924988-1G APOSOR924988-5G
1-Ethyl-1H-imidazole-2-carbaldehyde 95%
1-Ethyl-1H-imidazole-2-carbaldehyde
Formel: C₆H₈N₂O
Molecular Weight: 124,14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07380875
CAS-Nummer: 111851-98-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR