4-Propoxyphthalonitril 95%

Lieferant: Apollo Scientific

Synonyme: 1,2-Dicyano-4-propoxybenzene

OR925043-1G OR925043-5G
APOSOR925043-1GEA 284 EUR
APOSOR925043-1G APOSOR925043-5G
4-Propoxyphthalonitril 95%
4-Propoxyphthalonitril
Formel: C₁₁H₁₀N₂O
Molecular Weight: 186,21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00059102
CAS-Nummer: 106144-18-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR