2-Propyl-2-oxazolin 95%

Lieferant: Apollo Scientific

Synonyme: 2-Propyl-4,5-dihydrooxazole

OR925076-5G
APOSOR925076-5GEA 178 EUR
APOSOR925076-5G
2-Propyl-2-oxazolin 95%
2-Propyl-2-oxazolin
Formel: C₆H₁₁NO
Molecular Weight: 113,157 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02684776
CAS-Nummer: 4694-80-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR