α-Jonon 95%
Lieferant: Apollo Scientific
Synonyme:
(E)-4-(2,6,6-Trimethylcyclohex-2-en-1-yl)but-3-en-2-one
OR925128-500G
APOSOR925128-500GEA
116
EUR
APOSOR925128-500G
α-Jonon 95%
α-Jonon
Formel:
C₁₃H₂₀O Molecular Weight: 192,3 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001565 CAS-Nummer: 127-41-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...