3-Methoxybenzamid 95%

Lieferant: Apollo Scientific

Synonyme: m-Anisamid, 3-Methoxybenzoesäureamid

OR925166-25G
APOSOR925166-25GEA 79.9 EUR
APOSOR925166-25G
3-Methoxybenzamid 95%
3-Methoxybenzamid
Formel: C₈H₉NO₂
Molecular Weight: 151.16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00007986
CAS-Nummer: 5813-86-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR