3,5-Dimethylbenzylalkohol 97%

Lieferant: Apollo Scientific

Synonyme: (3,5-Dimethylphenyl)methanol, 5-(Hydroxymethyl)-m-xylol

OR925174-25G
APOSOR925174-25GEA 145 EUR
APOSOR925174-25G
3,5-Dimethylbenzylalkohol 97%
3,5-Dimethylbenzylalkohol
Formel: C₉H₁₂O
Molecular Weight: 136,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00004648
CAS-Nummer: 27129-87-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR