4'-Chlor-3'-methylacetophenon 99%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
1-(4-Chloro-3-methylphenyl)ethanone
OR925182-1G
OR925182-25G
OR925182-5G
APOSOR925182-1GEA
21.9
EUR
APOSOR925182-1G
APOSOR925182-25G
APOSOR925182-5G
4'-Chlor-3'-methylacetophenon 99%
4'-Chlor-3'-methylacetophenon
Formel:
C₉H₉ClO Molecular Weight: 168,62 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00045172 CAS-Nummer: 37074-39-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...