3-Methyl-2-buten-1-yl-acetat 97%
Lieferant: Apollo Scientific
Synonyme:
Acetic Acid Prenyl Ester, Acetic Acid 3-Methyl-2-butenyl Ester, 3-Methylbut-2-en-1-yl acetate, Prenyl Acetate
OR925190-100G
OR925190-1KG
OR925190-500G
APOSOR925190-100GEA
132
EUR
APOSOR925190-100G
APOSOR925190-1KG
APOSOR925190-500G
3-Methyl-2-buten-1-yl-acetat 97%
3-Methyl-2-buten-1-yl-acetat
Formel:
C₇H₁₂O₂ Molecular Weight: 128,17 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00036569 CAS-Nummer: 1191-16-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...