3-Ethyl-1-pentin-3-ol 95%
Lieferant: Apollo Scientific
Synonyme:
3-Ethyl-3-hydroxy-1-pentin, 3-Ethylpent-1-in-3-ol
OR925228-1G
OR925228-25G
OR925228-5G
APOSOR925228-1GEA
132
EUR
APOSOR925228-1G
APOSOR925228-25G
APOSOR925228-5G
3-Ethyl-1-pentin-3-ol 95%
3-Ethyl-1-pentin-3-ol
Formel:
C₇H₁₂O Molecular Weight: 112.17 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00014392 CAS-Nummer: 6285-06-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...