N-(2-Pyridinylmethyl)cyclohexanamin 95%

Lieferant: Apollo Scientific

Synonyme: N-(2-Pyridylmethyl)cyclohexylamine, N-Cyclohexyl-2-picolylamine

OR925247-1G OR925247-5G
APOSOR925247-1GEA 183 EUR
APOSOR925247-1G APOSOR925247-5G
N-(2-Pyridinylmethyl)cyclohexanamin 95%
2-(Cyclohexylaminomethyl)pyridine
Formel: C₁₂H₁₈N₂
Molecular Weight: 190,29 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01135534
CAS-Nummer: 68339-45-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR